Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boc-His Bom -OH, Advanced ChemTech

Catalog No. 01341127
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-341-127 25 g
01-341-126 5 g
01-341-128 1 g
01-341-129 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-341-127 Supplier Advanced Chem Tech Supplier No. BH231725G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 83468-83-1
Molecular Formula C19H25N3O5
Molecular Weight (g/mol) 375.43
InChI Key IEAOAWZLUGOPJX-UHFFFAOYNA-N
IUPAC Name 3-{1-[(benzyloxy)methyl]-1H-imidazol-4-yl}-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CN(COCC2=CC=CC=C2)C=N1)C(O)=O

Specifications

Color White to Off-White
Quantity 25 g
Grade Biochemical
Chemical Name or Material Boc-His Bom -OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.