Learn More
Apexbio Technology LLC Boc-His(Dnp)-OH 25024-53-7 25g
SDP
Catalog No. 5000088662
Encompass_Preferred
Boc-His Dnp -OH is a modified histidine derivative used in peptide synthesis It features a tert-butyloxycarbonyl (Boc) group protecting the -amino function and a dinitrophenyl (Dnp) group attached to the side-chain imidazole These modifications provide reversible protection of histidine s reactive sites and confer chromogenic properties facilitating spectrophotometric tracking of peptide synthesis or enzymatic cleavage processes Based on these properties Boc-His Dnp -OH is applied in biomedical research as a substrate analog for studying protease specificity enzyme-substrate interactions and catalytic mechanisms supporting biochemical characterization of enzyme kinetics and pharmaceutical peptide-target development
Catalog No. 50-000-88662 Supplier Apexbio Technology LLC Supplier No. A777525000
Boc-His Dnp -OH is a modified histidine derivative used in peptide synthesis It features a tert-butyloxycarbonyl (Boc) group protecting the -amino function and a dinitrophenyl (Dnp) group attached to the side-chain imidazole These modifications provide reversible protection of histidine s reactive sites and confer chromogenic properties facilitating spectrophotometric tracking of peptide synthesis or enzymatic cleavage processes Based on these properties Boc-His Dnp -OH is applied in biomedical research as a substrate analog for studying protease specificity enzyme-substrate interactions and catalytic mechanisms supporting biochemical characterization of enzyme kinetics and pharmaceutical peptide-target development