Learn More
Apexbio Technology LLC Boc-β-Homo-Pro-OH 26250-84-0 25g
SDP
Catalog No. 5000097288
Encompass_Preferred
Boc-beta-Homo-Pro-OH (CAS 26250-84-0) is a proline derivative featuring a -amino acid backbone with a tert-butyloxycarbonyl (Boc) protecting group Its distinctive -positioned cyclic structure imparts altered conformational flexibility compared to standard -amino acids Boc-beta-Homo-Pro-OH is widely utilized in peptide modification and structural studies to facilitate the synthesis of peptides with defined spatial configurations or constrained cyclic motifs It serves as an intermediate in the preparation and structural refinement of peptide-based molecules for pharmaceutical and biomedical research
Catalog No. 50-000-97288 Supplier Apexbio Technology LLC Supplier No. A666925000
Boc-beta-Homo-Pro-OH (CAS 26250-84-0) is a proline derivative featuring a -amino acid backbone with a tert-butyloxycarbonyl (Boc) protecting group Its distinctive -positioned cyclic structure imparts altered conformational flexibility compared to standard -amino acids Boc-beta-Homo-Pro-OH is widely utilized in peptide modification and structural studies to facilitate the synthesis of peptides with defined spatial configurations or constrained cyclic motifs It serves as an intermediate in the preparation and structural refinement of peptide-based molecules for pharmaceutical and biomedical research