Learn More
Apexbio Technology LLC Boc-β-Homo-Pro-OH 26250-84-0 5g
SDP
Catalog No. 5000093565
Encompass_Preferred
Boc-beta-Homo-Pro-OH (CAS 26250-84-0) is a proline derivative featuring a -amino acid backbone with a tert-butyloxycarbonyl (Boc) protecting group Its distinctive -positioned cyclic structure imparts altered conformational flexibility compared to standard -amino acids Boc-beta-Homo-Pro-OH is widely utilized in peptide modification and structural studies to facilitate the synthesis of peptides with defined spatial configurations or constrained cyclic motifs It serves as an intermediate in the preparation and structural refinement of peptide-based molecules for pharmaceutical and biomedical research
Catalog No. 50-000-93565 Supplier Apexbio Technology LLC Supplier No. A66695000
Boc-beta-Homo-Pro-OH (CAS 26250-84-0) is a proline derivative featuring a -amino acid backbone with a tert-butyloxycarbonyl (Boc) protecting group Its distinctive -positioned cyclic structure imparts altered conformational flexibility compared to standard -amino acids Boc-beta-Homo-Pro-OH is widely utilized in peptide modification and structural studies to facilitate the synthesis of peptides with defined spatial configurations or constrained cyclic motifs It serves as an intermediate in the preparation and structural refinement of peptide-based molecules for pharmaceutical and biomedical research