Learn More
Medchemexpress LLC Boc-Hyp-OEt | 37813-30-2 | MFCD09264314 | 99.7% | 259.30 g/mol | C12H21NO5 | 25 G
SDP
Catalog No. 5000208565
Encompass_Preferred
Boc-Hyp-OEt is the tert-butyloxycarbonyl (Boc) protected ethyl ester of hydroxyproline, provided as a high-purity building block for peptide synthesis and organic chemistry research. It is supplied with analytical documentation to support research applications.
  • High purity per certificate of analysis.
  • Suitable for use as a protected amino acid in peptide synthesis.
  • Provided as a viscous, colorless to light yellow liquid for ease of handling.
  • Available in common laboratory pack sizes for small-scale synthesis.
  • Analytical documentation available, including COA and safety data sheet.
Catalog No. 50-002-08565 Supplier Medchemexpress LLC Supplier No. 25GHYW052529
Boc-Hyp-OEt is the tert-butyloxycarbonyl (Boc) protected ethyl ester of hydroxyproline, provided as a high-purity building block for peptide synthesis and organic chemistry research. It is supplied with analytical documentation to support research applications.
  • High purity per certificate of analysis.
  • Suitable for use as a protected amino acid in peptide synthesis.
  • Provided as a viscous, colorless to light yellow liquid for ease of handling.
  • Available in common laboratory pack sizes for small-scale synthesis.
  • Analytical documentation available, including COA and safety data sheet.