Learn More
Medchemexpress LLC Boc-Hyp-OH | 13726-69-7 | 99.8% | 231.25 g/mol | C10H17NO5 | 5 G
SDP
Catalog No. 5000202742
Encompass_Preferred
Boc-Hyp-OH is the tert-butyloxycarbonyl (Boc) protected form of hydroxyproline, used as a protected amino-acid building block for peptide synthesis and as a linker component in antibody-drug conjugate (ADC) and PROTAC chemistry.
  • Protected amino acid for solid-phase and solution-phase peptide synthesis.
  • Suitable as a linker component in ADC and PROTAC synthesis.
  • High purity for research applications.
  • Available in multiple pack sizes for research-scale use.
Catalog No. 50-002-02742 Supplier Medchemexpress LLC Supplier No. HYI07815G
Boc-Hyp-OH is the tert-butyloxycarbonyl (Boc) protected form of hydroxyproline, used as a protected amino-acid building block for peptide synthesis and as a linker component in antibody-drug conjugate (ADC) and PROTAC chemistry.
  • Protected amino acid for solid-phase and solution-phase peptide synthesis.
  • Suitable as a linker component in ADC and PROTAC synthesis.
  • High purity for research applications.
  • Available in multiple pack sizes for research-scale use.