Learn More
Medchemexpress LLC Boc-Ile-Glu-Gly-Arg-AMC (IEGR-AMC) | 65147-06-0 | 99.2% | 730.81 | 10 MG
SDP
Catalog No. 5000422888
Encompass_Preferred
Boc-Ile-Glu-Gly-Arg-AMC (IEGR-AMC) is a fluorogenic peptide substrate specific for activated factor X (FXa), utilized in the determination of Factor VIII. This compound presents as a white to off-white solid.
  • Fluorogenic peptide substrate
  • Specific for activated factor X (FXa)
  • Used for Factor VIII determination
  • White to off-white solid appearance
  • Ships at room temperature in continental US
  • Store sealed, away from moisture
  • Powder storage: -80°C for 2 years, -20°C for 1 year
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-004-22888 Supplier Medchemexpress LLC Supplier No. HYP200810MG
Boc-Ile-Glu-Gly-Arg-AMC (IEGR-AMC) is a fluorogenic peptide substrate specific for activated factor X (FXa), utilized in the determination of Factor VIII. This compound presents as a white to off-white solid.
  • Fluorogenic peptide substrate
  • Specific for activated factor X (FXa)
  • Used for Factor VIII determination
  • White to off-white solid appearance
  • Ships at room temperature in continental US
  • Store sealed, away from moisture
  • Powder storage: -80°C for 2 years, -20°C for 1 year
  • In solvent storage: -80°C for 6 months, -20°C for 1 month