Learn More
Medchemexpress LLC Boc-L-Dap-OH | 73259-81-1 | 204.23 | 25 G
SDP
Catalog No. 5000448238
Encompass_Preferred
Boc-L-Dap-OH is a Boc-protected derivative of L-2,3-diaminopropionic acid. It functions as an important chiral amino acid derivative and can be synthesized via the Hofmann rearrangement of Boc-L-asparagine. This compound serves as a starting material for the preparation of BIM analogs.
  • Boc-protected derivative of L-2,3-diaminopropionic acid
  • Important chiral amino acid derivative
  • Synthesized via Hofmann rearrangement
  • Starting material for BIM analogs
  • Appears as a white to off-white solid
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage for in solvent: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-004-48238 Supplier Medchemexpress LLC Supplier No. 25GHYW008325
Boc-L-Dap-OH is a Boc-protected derivative of L-2,3-diaminopropionic acid. It functions as an important chiral amino acid derivative and can be synthesized via the Hofmann rearrangement of Boc-L-asparagine. This compound serves as a starting material for the preparation of BIM analogs.
  • Boc-protected derivative of L-2,3-diaminopropionic acid
  • Important chiral amino acid derivative
  • Synthesized via Hofmann rearrangement
  • Starting material for BIM analogs
  • Appears as a white to off-white solid
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage for in solvent: -80°C for 6 months, -20°C for 1 month