Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical Boc-L-Lys(Fmoc)-OMe | 25 g | CAS 133628-28-1 | MDL MFCD00236856

Catalog No. 502352676
Encompass

Boc-L-Lys(Fmoc)-OMe is a Amino Acid reagent (Subcategory: Lys) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 133628-28-1
- MDL: MFCD00236856
- InChIKey: NUSZDVABASFMMF-QHCPKHFHSA-N
- Molecular Weight: 482.577
- Molecular Formula: C27H34N2O6
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: methyl N6-(((9H-fluoren-9-yl)methoxy)carbonyl)-N2-(tert-butoxycarbonyl)-L-lysinate
- SMILES: CC(OC(N[C@@H](CCCCNC(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O)C(OC)=O)=O)(C)C

Catalog No. 50-235-2676 Supplier STA Pharmaceutical Supplier No. STAAA1058025GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-L-Lys(Fmoc)-OMe is a Amino Acid reagent (Subcategory: Lys) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 133628-28-1
- MDL: MFCD00236856
- InChIKey: NUSZDVABASFMMF-QHCPKHFHSA-N
- Molecular Weight: 482.577
- Molecular Formula: C27H34N2O6
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: methyl N6-(((9H-fluoren-9-yl)methoxy)carbonyl)-N2-(tert-butoxycarbonyl)-L-lysinate
- SMILES: CC(OC(N[C@@H](CCCCNC(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O)C(OC)=O)=O)(C)C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.