Learn More
AnaSpec Boc-L-propargylglycine-DCHA, | MW 213.2-181.3 | M.F. C10H15NO4 (1 g)
Supplier:  AnaSpec AS26264B1
Encompass
Boc-L-propargylglycine-DCHA, ; MW 213.2-181.3; M.F. C10H15NO4 (1 g) An unusual amino acid used in Solid Phase Peptide Synthesis (SPPS) as: A) a precursor to the tritium-induced Norvaline peptides,1 B) in the synthesis of branched peptides using a palladium-catalyzed coupling reaction,2 and C) as a reagent in the dual-catalyst Sonogashira coupling reaction.3
Catalog No. 50351096