Learn More
Bachem Boc-L-thiazolidine-4-carboxylic acid
SDP
Catalog No. 50258458
Encompass_Preferred
Boc-L-thiazolidine-4-carboxylic acid, 51077-16-8, C9H15NO4S, Mr 233.29, 5g. Boc-Thz-OH, Boc-L-thioproline. Boc-Thz-OH has been applied as a masked N-terminal Cys in native chemical ligation. The thioaminal can be cleaved with methoxyamine · HCl.
Catalog No. 50-258-458 Supplier Bachem Supplier No. 4029316.0005
Add to Cart
Add to Cart
Boc-L-thiazolidine-4-carboxylic acid, 51077-16-8, C9H15NO4S, Mr 233.29, 5g. Boc-Thz-OH, Boc-L-thioproline. Boc-Thz-OH has been applied as a masked N-terminal Cys in native chemical ligation. The thioaminal can be cleaved with methoxyamine · HCl.