Learn More
Medchemexpress LLC Boc-L-Valine-d8 | 153568-33-3 | 225.31 | 10 MG
SDP
Catalog No. 5000445624
Encompass_Preferred
Boc-L-Valine-d8 is the deuterium labeled Boc-L-Valine, intended for research use only. This compound serves as a tracer and internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS. Deuteration, which involves incorporating stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, is utilized for quantitation during drug development and can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements
  • Used for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-45624 Supplier Medchemexpress LLC Supplier No. HYW002300S10MG
Boc-L-Valine-d8 is the deuterium labeled Boc-L-Valine, intended for research use only. This compound serves as a tracer and internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS. Deuteration, which involves incorporating stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, is utilized for quantitation during drug development and can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements
  • Used for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs