Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BOC-L-Valine hydroxysuccinimide ester, 98%

Catalog No. AC440920050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC440920050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC440920050 Supplier Thermo Scientific Chemicals Supplier No. 440920050
Add to Cart

CAS: 3392-12-9 | C14H22N2O6 | 314.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3392-12-9
Molecular Formula C14H22N2O6
Molecular Weight (g/mol) 314.34
MDL Number MFCD00037906
InChI Key POBDBYGSGKMZPH-NSHDSACASA-N
Synonym boc-val-osu, boc-l-valine hydroxysuccinimide ester, boc-l-val-osu, tert-butyl s-1-2,5-dioxopyrrolidin-1-yl oxy carbonyl-2-methylpropyl carbamate, 2,5-dioxopyrrolidin-1-yl n-tert-butoxycarbonyl-l-valinate, bocvalosu, t-boc-val-osu, pubchem20978, s-2,5-dioxopyrrolidin-1-yl 2-tert-butoxycarbonyl amino-3-methylbutanoate, boc-val-osu n
PubChem CID 98768
IUPAC Name 2,5-dioxopyrrolidin-1-yl (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-methylbutanoate
SMILES CC(C)[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O

Specifications

Melting Point 126°C to 129°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 5 g
Specific Rotation -37.2
Formula Weight 314.34
Percent Purity 98%
Physical Form Powder
Chemical Name or Material BOC-L-Valine hydroxysuccinimide ester
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 222-236-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.