Learn More
AdipoGen Boc-Leu-Arg-Arg-AMC
SDP
Catalog No. 502562448
Encompass_Preferred
Chemical. CAS 109358-46-5 free base. Formula C33H52N10O7 . C2H4O2. MW 700.8 . 60.1. Fluorogenic tri-peptide substrate for measuring the trypsin-like peptidase activity of the 20S proteasome. Substrate for the Kex2 endopeptidase from S. Cervisiae, which has substrate specificity toward the C-terminal side of LR, PR and RR sequences. Can be cleaved by Kallikrein 5 with a P1 site preference for basic residues R and K. Excitation 380nm. Emission 460nm. This substrate is useful for inhibitor screening and kinetic analysis.
Catalog No. 50-256-2448 Supplier AdipoGen Supplier No. AGCP30014M005
Chemical. CAS 109358-46-5 free base. Formula C33H52N10O7 . C2H4O2. MW 700.8 . 60.1. Fluorogenic tri-peptide substrate for measuring the trypsin-like peptidase activity of the 20S proteasome. Substrate for the Kex2 endopeptidase from S. Cervisiae, which has substrate specificity toward the C-terminal side of LR, PR and RR sequences. Can be cleaved by Kallikrein 5 with a P1 site preference for basic residues R and K. Excitation 380nm. Emission 460nm. This substrate is useful for inhibitor screening and kinetic analysis.