Learn More
Enzo Life Sciences Boc-Leu-Arg-Arg-AMC (5mg)
SDP
Catalog No. 502010339
Encompass_Preferred
A substrate for the Kex2 endopeptidase from S. cerevisiae. This peptide is also a substrate for kallikrein 5 (KLK5), and a candidate substrate for the investigation of proteasome subunit function. Alternative name: Boc-LRR-AMC. Purity: ≥97% (HPLC). Solubility: Soluble in water (50mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0339 Supplier Enzo Life Sciences Supplier No. BMLBW85150005
A substrate for the Kex2 endopeptidase from S. cerevisiae. This peptide is also a substrate for kallikrein 5 (KLK5), and a candidate substrate for the investigation of proteasome subunit function. Alternative name: Boc-LRR-AMC. Purity: ≥97% (HPLC). Solubility: Soluble in water (50mg/ml). Long Term Storage: -20°C.