Learn More
Medchemexpress LLC Boc-Leu-Gly-Arg-AMC | 65147-09-3 | 98.3% | 601.69 | 5 MG
SDP
Catalog No. 5000422216
Encompass_Preferred
Boc-Leu-Gly-Arg-AMC is a fluorogenic AMC substrate primarily used for convertases. This compound is suitable for use in enzymatic assays. Research has shown its application in developing enzymatic assays to detect proteolytic enzymes, such as those capable of generating sodefrin, by measuring the liberation of 7-amino-4-methylcoumarin. The optimal activity for cleaving Boc-Leu-Gly-Arg-AMC in certain extracts has been observed at pH 9.0 and 45 °C.
  • Fluorogenic AMC substrate for convertases.
  • Can be used in enzymatic assays.
  • Suitable for demonstrating the presence of proteolytic enzymes.
Catalog No. 50-004-22216 Supplier Medchemexpress LLC Supplier No. HYP22375MG
Boc-Leu-Gly-Arg-AMC is a fluorogenic AMC substrate primarily used for convertases. This compound is suitable for use in enzymatic assays. Research has shown its application in developing enzymatic assays to detect proteolytic enzymes, such as those capable of generating sodefrin, by measuring the liberation of 7-amino-4-methylcoumarin. The optimal activity for cleaving Boc-Leu-Gly-Arg-AMC in certain extracts has been observed at pH 9.0 and 45 °C.
  • Fluorogenic AMC substrate for convertases.
  • Can be used in enzymatic assays.
  • Suitable for demonstrating the presence of proteolytic enzymes.