Learn More
Medchemexpress LLC Boc-leucinol | 82010-31-9 | MFCD00076950 | 95.0% | 217.31 g·mol⁻¹ | C11H23NO3 | 100 G
SDP
Catalog No. 5000304094
Encompass_Preferred
Boc-leucinol is a Boc-protected derivative of the amino acid leucine in which the carboxyl group has been reduced to the corresponding alcohol. It is a chiral, primary alcohol reagent commonly used as a synthetic intermediate and building block in peptide chemistry and small-molecule synthesis, suitable for introducing protected amino alcohol functionality.
  • Boc-protected amino alcohol functionality for synthetic versatility.
  • Retains chiral center for stereoselective synthesis.
  • Suitable as a peptide intermediate or building block in medicinal chemistry.
  • Viscous liquid form simplifies handling and dispensing.
  • Compatible with common protecting group manipulations and coupling reactions.
Catalog No. 50-003-04094 Supplier Medchemexpress LLC Supplier No. HYW014097100G
Boc-leucinol is a Boc-protected derivative of the amino acid leucine in which the carboxyl group has been reduced to the corresponding alcohol. It is a chiral, primary alcohol reagent commonly used as a synthetic intermediate and building block in peptide chemistry and small-molecule synthesis, suitable for introducing protected amino alcohol functionality.
  • Boc-protected amino alcohol functionality for synthetic versatility.
  • Retains chiral center for stereoselective synthesis.
  • Suitable as a peptide intermediate or building block in medicinal chemistry.
  • Viscous liquid form simplifies handling and dispensing.
  • Compatible with common protecting group manipulations and coupling reactions.