Learn More
Cayman Chemical Boc-LGR-pNA (acetate), Purity: ≥98%, CAS: 176786-91-7, 10 mg
SDP
Catalog No. 502911280
Encompass_Preferred
A chromogenic substrate for endotoxins; enzymatic cleavage releases pNA which can be quantified by colorimetric detection at 405 nm as a measure of endotoxin levels
Catalog No. 50-291-1280 Supplier Cayman Chemical Supplier No. 3517610 MG
A chromogenic substrate for endotoxins; enzymatic cleavage releases pNA which can be quantified by colorimetric detection at 405 nm as a measure of endotoxin levels