Learn More
Bachem Boc-Leu-Arg-Arg-AMC · 2 HCl
SDP
Catalog No. 501947051
Encompass_Preferred
Boc-Leu-Arg-Arg-AMC · 2 HCl, 109358-46-5, C33H52N10O7 · 2 HCl, Mr 773.76, 50mg. Boc-LLR-AMC is a good substrate for the Kex2 endoprotease (kexin, previously known as proteinase yscF), a membrane-bound, calcium-dependent serine protease from yeast a-cells. Boc-LRR-AMC is also used for determining the trypsin-like activity of the proteasome.
Catalog No. 50-194-7051 Supplier Bachem Supplier No. 4017087.0050
Boc-Leu-Arg-Arg-AMC · 2 HCl, 109358-46-5, C33H52N10O7 · 2 HCl, Mr 773.76, 50mg. Boc-LLR-AMC is a good substrate for the Kex2 endoprotease (kexin, previously known as proteinase yscF), a membrane-bound, calcium-dependent serine protease from yeast a-cells. Boc-LRR-AMC is also used for determining the trypsin-like activity of the proteasome.