Learn More
Medchemexpress LLC Boc-LRR-AMC | 25MG
SDP
Catalog No. 5000227089
Encompass_Preferred
Boc-LRR-AMC (Boc-Leu-Arg-Arg-AMC) is a fluorogenic substrate which can be utilized for detecting the trypsin-like activity of the 26S proteasome or the 20S proteasome core[1]
Catalog No. 50-002-27089 Supplier Medchemexpress LLC Supplier No. HY11539125MG
Boc-LRR-AMC (Boc-Leu-Arg-Arg-AMC) is a fluorogenic substrate which can be utilized for detecting the trypsin-like activity of the 26S proteasome or the 20S proteasome core[1]