Learn More
Medchemexpress LLC Boc-LRR-AMC | 700.83 g/mol | C33H52N10O7 | 10MG
SDP
Catalog No. 5000307633
Encompass_Preferred
Boc-LRR-AMC (Boc-Leu-Arg-Arg-AMC) is a fluorogenic substrate which can be utilized for detecting the trypsin-like activity of the 26S proteasome or the 20S proteasome core[1]
Catalog No. 50-003-07633 Supplier Medchemexpress LLC Supplier No. HY11539110MG
Boc-LRR-AMC (Boc-Leu-Arg-Arg-AMC) is a fluorogenic substrate which can be utilized for detecting the trypsin-like activity of the 26S proteasome or the 20S proteasome core[1]