Learn More
Bachem Boc-Leu-Ser-Thr-Arg-AMC
SDP
Catalog No. 501947070
Encompass_Preferred
Boc-Leu-Ser-Thr-Arg-AMC Trifluoroacetate, 73554-93-5, C34H52N8O10, Mr 732.83, 25mg. Boc-LSTR-AMC. Boc-LSTR-AMC is a good substrate for the activated protein C (Km = 3.3 · 10-4 M, kcat = 8.4 s-1). The fluorogenic substrate is often used for assaying the tryptic activity of the proteasome.
Catalog No. 50-194-7070 Supplier Bachem Supplier No. 4044927.0025
Add to Cart
Add to Cart
Boc-Leu-Ser-Thr-Arg-AMC Trifluoroacetate, 73554-93-5, C34H52N8O10, Mr 732.83, 25mg. Boc-LSTR-AMC. Boc-LSTR-AMC is a good substrate for the activated protein C (Km = 3.3 · 10-4 M, kcat = 8.4 s-1). The fluorogenic substrate is often used for assaying the tryptic activity of the proteasome.