Learn More
Cayman Chemical Boc-Lys(Ac)-AMC, Purity: ≥98%, CAS: 233691-67-3, 10 mg
SDP
Catalog No. 502988344
Encompass_Preferred
A fluorogenic HDAC substrate; ex/em = 340-360/440-460 nm, respectively; following deacetylation by an HDAC and trypsin cleavage, AMC is released and its fluorescence can be used to quantify HDAC activity
Catalog No. 50-298-8344 Supplier Cayman Chemical Supplier No. 2968210 MG
A fluorogenic HDAC substrate; ex/em = 340-360/440-460 nm, respectively; following deacetylation by an HDAC and trypsin cleavage, AMC is released and its fluorescence can be used to quantify HDAC activity