Learn More
Apexbio Technology LLC Boc-Lys(Ac)-AMC 233691-67-3 250mg
SDP
Catalog No. 5000096190
Encompass_Preferred
Boc-Lys Ac -AMC (CAS 233691-67-3) is a fluorescently labeled peptide substrate designed for analyzing enzymatic activity of histone deacetylases (HDACs) It enables monitoring of HDAC-mediated deacetylation reactions Boc-Lys Ac -AMC exerts its biological activity primarily through acting as a fluorogenic substrate for HDACs where deacetylation leads to subsequent fluorescence release following proteolytic cleavage Based on these properties Boc-Lys Ac -AMC holds research potential in biochemical assays and drug screening studies for characterizing HDAC enzyme kinetics and assessing inhibitor potency related to epigenetic modulation
Catalog No. 50-000-96190 Supplier Apexbio Technology LLC Supplier No. A8713250
Boc-Lys Ac -AMC (CAS 233691-67-3) is a fluorescently labeled peptide substrate designed for analyzing enzymatic activity of histone deacetylases (HDACs) It enables monitoring of HDAC-mediated deacetylation reactions Boc-Lys Ac -AMC exerts its biological activity primarily through acting as a fluorogenic substrate for HDACs where deacetylation leads to subsequent fluorescence release following proteolytic cleavage Based on these properties Boc-Lys Ac -AMC holds research potential in biochemical assays and drug screening studies for characterizing HDAC enzyme kinetics and assessing inhibitor potency related to epigenetic modulation