Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-Lys(Ac)-AMC | 233691-67-3 | 98.59% | 445.51 | 1 ML
SDP

Catalog No. 5000348596
Encompass_Preferred

Boc-Lys(Ac)-AMC is a cell-permeable fluorometric HDAC substrate with excitation/emission wavelengths of 355 nm/460 nm. It is intended for research use only and not for sale to patients. This product is used in tissue culture-based HDAC assays.

  • Cell-permeable fluorometric HDAC substrate
  • Excitation/emission wavelengths of 355 nm/460 nm
  • Store solid at -20°C, sealed, away from moisture and light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture and light

Catalog No. 50-003-48596 Supplier Medchemexpress LLC Supplier No. HY13815910MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-Lys(Ac)-AMC is a cell-permeable fluorometric HDAC substrate with excitation/emission wavelengths of 355 nm/460 nm. It is intended for research use only and not for sale to patients. This product is used in tissue culture-based HDAC assays.

  • Cell-permeable fluorometric HDAC substrate
  • Excitation/emission wavelengths of 355 nm/460 nm
  • Store solid at -20°C, sealed, away from moisture and light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture and light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.