Learn More
Medchemexpress LLC Boc-Lys(Ac)-AMC | 233691-67-3 | 98.6% | 445.51 | 100 MG
SDP
Catalog No. 5000350016
Encompass_Preferred
Boc-Lys(Ac)-AMC is a cell-permeable fluorometric substrate utilized in assays for Histone Deacetylase (HDAC) activity, with excitation and emission wavelengths at 355 nm and 460 nm, respectively. It is suitable for research purposes only.
  • Cell-permeable
  • Fluorometric substrate
  • Used for measuring HDAC inhibition
  • Applicable in tissue culture-based HDAC assays
Catalog No. 50-003-50016 Supplier Medchemexpress LLC Supplier No. HY138159100MG
Boc-Lys(Ac)-AMC is a cell-permeable fluorometric substrate utilized in assays for Histone Deacetylase (HDAC) activity, with excitation and emission wavelengths at 355 nm and 460 nm, respectively. It is suitable for research purposes only.
  • Cell-permeable
  • Fluorometric substrate
  • Used for measuring HDAC inhibition
  • Applicable in tissue culture-based HDAC assays