Learn More
Medchemexpress LLC Boc-Lys(Ac)-AMC | 233691-67-3 | 98.6% | 445.51 | 25 MG
SDP
Catalog No. 5000348610
Encompass_Preferred
Boc-Lys(Ac)-AMC is a cell-permeable fluorometric HDAC substrate with excitation/emission wavelengths of 355 nm/460 nm.
  • Cell-permeable fluorometric HDAC substrate
  • Excitation/Emission = 355 nm/460 nm
Catalog No. 50-003-48610 Supplier Medchemexpress LLC Supplier No. 25MGHY138159
Boc-Lys(Ac)-AMC is a cell-permeable fluorometric HDAC substrate with excitation/emission wavelengths of 355 nm/460 nm.
  • Cell-permeable fluorometric HDAC substrate
  • Excitation/Emission = 355 nm/460 nm