Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-Lys(Ac)-AMC | 233691-67-3 | MFCD02683946 | 99.5% | C23H31N3O6 | 10MG
SDP

Catalog No. 5000261968
Encompass_Preferred

Boc-Lys(Ac)-AMC is a cell-permeable fluorogenic substrate for histone deacetylases (HDACs). Upon enzymatic deacetylation and subsequent proteolytic cleavage, it releases 7-amino-4-methylcoumarin (AMC), producing a fluorescent signal suitable for quantitative fluorometric assays and inhibitor screening (Ex/Em = 355 nm/460 nm).

  • Cell-permeable fluorogenic histone deacetylase substrate.
  • Produces fluorescent AMC upon deacetylation and proteolytic cleavage.
  • Excitation/emission: 355 nm/460 nm for sensitive detection.
  • High purity suitable for enzymatic activity and inhibitor assays.
  • Soluble in DMSO (≈100 mg/mL); sonication may be required.
  • Store sealed, away from moisture and light to maintain stability.

Catalog No. 50-002-61968 Supplier Medchemexpress LLC Supplier No. HY13815910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-Lys(Ac)-AMC is a cell-permeable fluorogenic substrate for histone deacetylases (HDACs). Upon enzymatic deacetylation and subsequent proteolytic cleavage, it releases 7-amino-4-methylcoumarin (AMC), producing a fluorescent signal suitable for quantitative fluorometric assays and inhibitor screening (Ex/Em = 355 nm/460 nm).

  • Cell-permeable fluorogenic histone deacetylase substrate.
  • Produces fluorescent AMC upon deacetylation and proteolytic cleavage.
  • Excitation/emission: 355 nm/460 nm for sensitive detection.
  • High purity suitable for enzymatic activity and inhibitor assays.
  • Soluble in DMSO (≈100 mg/mL); sonication may be required.
  • Store sealed, away from moisture and light to maintain stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.