Learn More
AnaSpec BOC-LYS(N3)-OH
Supplier:  AnaSpec AS53100B1
Encompass
Boc-Lys(N3)-OH[Boc-azidolysine; Boc-lys(azide); Boc-lysine azide] ; MW 272.3; M.F. C11H20N4O4 (1 g) An unusual amino acid used in Solid Phase Peptide Synthesis (SPPS) for the synthesis of side-chain modified peptides and proteins.1 The side-chain azido (N3) group is stable in trifluoroacetic acid (TFA) or piperidine (Pip), two reagents commonly used in SPPS. The azido group can be readily converted to an amine on the solid phase or in solution via the Staudinger reduction.2
Catalog No. 50352209