Learn More
Apexbio Technology LLC Boc-Lys-OH 13734-28-6 25g
SDP
Catalog No. 5000092109
Encompass_Preferred
Boc-Lys-OH (13734-28-6) is a lysine derivative protected with a tert-butyloxycarbonyl (Boc) group at the alpha-amino position enabling selective conjugation through the lysine side-chain amine Its chemical characteristics facilitate the targeted linkage of functional groups to peptides and proteins Boc-Lys-OH serves primarily as an intermediate in peptide synthesis allowing controlled introduction of molecular labels tags fluorophores or drug moieties Based on these properties Boc-Lys-OH holds research potential in applications such as antibody-drug conjugate (ADC) construction fluorescent peptide probes protein-protein interaction studies cellular signaling investigation and the development of targeted delivery systems
Catalog No. 50-000-92109 Supplier Apexbio Technology LLC Supplier No. A778625000
Boc-Lys-OH (13734-28-6) is a lysine derivative protected with a tert-butyloxycarbonyl (Boc) group at the alpha-amino position enabling selective conjugation through the lysine side-chain amine Its chemical characteristics facilitate the targeted linkage of functional groups to peptides and proteins Boc-Lys-OH serves primarily as an intermediate in peptide synthesis allowing controlled introduction of molecular labels tags fluorophores or drug moieties Based on these properties Boc-Lys-OH holds research potential in applications such as antibody-drug conjugate (ADC) construction fluorescent peptide probes protein-protein interaction studies cellular signaling investigation and the development of targeted delivery systems