Learn More
Bachem Boc-Lys(Tfa)-AMC
SDP
Catalog No. 50260364
Encompass_Preferred
Boc-Lys(Tfa)-AMC, 97885-44-4, C23H28F3N3O6, Mr 499.49, 50mg. Fluorogenic substrate for assaying histone deacetylase (HDAC) 4, 5 and 7 activity in a protease-coupled assay. Suitable substrate for HDAC 8. HDAC cleavage yields Boc-Lys-AMC.
Catalog No. 50-260-364 Supplier Bachem Supplier No. 4060676.0050
Boc-Lys(Tfa)-AMC, 97885-44-4, C23H28F3N3O6, Mr 499.49, 50mg. Fluorogenic substrate for assaying histone deacetylase (HDAC) 4, 5 and 7 activity in a protease-coupled assay. Suitable substrate for HDAC 8. HDAC cleavage yields Boc-Lys-AMC.