Learn More
Apexbio Technology LLC Boc-Lys(Ac)-AMC 233691-67-3 50mg
SDP
Catalog No. 502240092
Encompass_Preferred
Boc-Lys Ac -AMC (CAS 233691-67-3) is a fluorescently labeled peptide substrate designed for analyzing enzymatic activity of histone deacetylases (HDACs) It enables monitoring of HDAC-mediated deacetylation reactions Boc-Lys Ac -AMC exerts its biological activity primarily through acting as a fluorogenic substrate for HDACs where deacetylation leads to subsequent fluorescence release following proteolytic cleavage Based on these properties Boc-Lys Ac -AMC holds research potential in biochemical assays and drug screening studies for characterizing HDAC enzyme kinetics and assessing inhibitor potency related to epigenetic modulation
This item is not returnable. View return policy
Catalog No. 50-224-0092 Supplier Apexbio Technology LLC Supplier No. A871350
Boc-Lys Ac -AMC (CAS 233691-67-3) is a fluorescently labeled peptide substrate designed for analyzing enzymatic activity of histone deacetylases (HDACs) It enables monitoring of HDAC-mediated deacetylation reactions Boc-Lys Ac -AMC exerts its biological activity primarily through acting as a fluorogenic substrate for HDACs where deacetylation leads to subsequent fluorescence release following proteolytic cleavage Based on these properties Boc-Lys Ac -AMC holds research potential in biochemical assays and drug screening studies for characterizing HDAC enzyme kinetics and assessing inhibitor potency related to epigenetic modulation