Learn More
Medchemexpress LLC {Boc}-Met-Leu-Phe | 67247-12-5 | MFCD00037435 | 99.9% | 509.66 g/mol | C25H39N3O6S | 5 MG
SDP
Catalog No. 5000248252
Encompass_Preferred
Boc-MLF (Boc-Met-Leu-Phe-OH) is a Boc-protected tripeptide that acts as a formyl peptide receptor (FPR) antagonist. It reduces fMLF-induced superoxide production with a reported IC50 of 0.63 μM and is supplied as a purified powder for research use. The compound is readily soluble in DMSO and can be formulated for in vivo dosing using common co-solvents.
Catalog No. 50-002-48252 Supplier Medchemexpress LLC Supplier No. HY1034735MG
Boc-MLF (Boc-Met-Leu-Phe-OH) is a Boc-protected tripeptide that acts as a formyl peptide receptor (FPR) antagonist. It reduces fMLF-induced superoxide production with a reported IC50 of 0.63 μM and is supplied as a purified powder for research use. The compound is readily soluble in DMSO and can be formulated for in vivo dosing using common co-solvents.