Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-Met-Leu-Phe-OH | 67247-12-5 | MFCD00037435 | 99.9% | 509.66 | C25H39N3O6S | 25 MG
SDP

Catalog No. 5000249270
Encompass_Preferred

Boc-Met-Leu-Phe-OH is a formyl peptide receptor (FPR) antagonist that reduces superoxide production induced by fMLF, with an IC50 of 0.63 μM. It is supplied as a high-purity peptide for research use in studies of FPR signaling and neutrophil activation.

  • Reduces fMLF-induced superoxide production (IC50 = 0.63 μM)
  • High purity (99.9%)
  • Molecular weight 509.66 g/mol
  • Molecular formula C25H39N3O6S
  • Available in small mg quantities for lab-scale studies

Catalog No. 50-002-49270 Supplier Medchemexpress LLC Supplier No. HY10347325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-Met-Leu-Phe-OH is a formyl peptide receptor (FPR) antagonist that reduces superoxide production induced by fMLF, with an IC50 of 0.63 μM. It is supplied as a high-purity peptide for research use in studies of FPR signaling and neutrophil activation.

  • Reduces fMLF-induced superoxide production (IC50 = 0.63 μM)
  • High purity (99.9%)
  • Molecular weight 509.66 g/mol
  • Molecular formula C25H39N3O6S
  • Available in small mg quantities for lab-scale studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.