Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-Met-Leu-Phe-OH | 67247-12-5 | MFCD00037435 | 99.9% | 509.66 g/mol | C25H39N3O6S | 50 MG
SDP

Catalog No. 5000246125
Encompass_Preferred

Boc-MLF (Boc-Met-Leu-Phe-OH) is a Boc-protected tripeptide that functions as a formyl peptide receptor (FPR) antagonist. It reduces superoxide production induced by fMLF with an IC50 of 0.63 μM and is supplied as a high-purity research reagent for biochemical and cellular assays.

  • High purity (99.93%).
  • Chemical formula C25H39N3O6S.
  • Molecular weight 509.66 g/mol.
  • Available in milligram pack sizes for small-scale assays.
  • Active as an FPR antagonist; reduces fMLF-induced superoxide production (IC50 = 0.63 μM).

Catalog No. 50-002-46125 Supplier Medchemexpress LLC Supplier No. HY10347350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-MLF (Boc-Met-Leu-Phe-OH) is a Boc-protected tripeptide that functions as a formyl peptide receptor (FPR) antagonist. It reduces superoxide production induced by fMLF with an IC50 of 0.63 μM and is supplied as a high-purity research reagent for biochemical and cellular assays.

  • High purity (99.93%).
  • Chemical formula C25H39N3O6S.
  • Molecular weight 509.66 g/mol.
  • Available in milligram pack sizes for small-scale assays.
  • Active as an FPR antagonist; reduces fMLF-induced superoxide production (IC50 = 0.63 μM).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.