Learn More
Medchemexpress LLC Boc-Met-Leu-Phe-OH trifluoroacetate | 00-00-0 | MFCD00037435 | 99.5% | 623.68 g·mol⁻¹ | C27H40F3N3O8S | 10 MG
SDP
Catalog No. 5000249228
Encompass_Preferred
Boc-MLF TFA is a protected tripeptide (Boc-Met-Leu-Phe-OH trifluoroacetate) used as a formyl peptide receptor antagonist in biochemical and cell-signaling research. It can inhibit signaling through formyl peptide receptor-like 1 (FPRL1) at higher concentrations. The material is supplied as a solid and in pre-made DMSO solutions and is for research use only.
  • Formyl peptide receptor antagonist peptide
  • Inhibits FPRL1 signaling at higher concentrations
  • Reported purity approximately 99.5% by manufacturer
  • Available as solid and as pre-dissolved DMSO solutions
  • Molecular weight approximately 623.68 g·mol⁻¹ (TFA salt)
  • Intended for research use only; not for clinical or patient use
Catalog No. 50-002-49228 Supplier Medchemexpress LLC Supplier No. HY103473A10MG
Boc-MLF TFA is a protected tripeptide (Boc-Met-Leu-Phe-OH trifluoroacetate) used as a formyl peptide receptor antagonist in biochemical and cell-signaling research. It can inhibit signaling through formyl peptide receptor-like 1 (FPRL1) at higher concentrations. The material is supplied as a solid and in pre-made DMSO solutions and is for research use only.
  • Formyl peptide receptor antagonist peptide
  • Inhibits FPRL1 signaling at higher concentrations
  • Reported purity approximately 99.5% by manufacturer
  • Available as solid and as pre-dissolved DMSO solutions
  • Molecular weight approximately 623.68 g·mol⁻¹ (TFA salt)
  • Intended for research use only; not for clinical or patient use