Learn More
Medchemexpress LLC Boc-Met-Leu-Phe (trifluoroacetate) | 00-00-0 | MFCD00037435 | 99.5% | 623.68 g/mol | C27H40F3N3O8S | 1 ML
SDP
Catalog No. 5000248919
Encompass_Preferred
Boc-MLF TFA is a Boc-protected tripeptide (Boc-Met-Leu-Phe as the trifluoroacetate salt) used as a formyl peptide receptor (FPR) antagonist for receptor pharmacology and cell signaling studies. It is supplied as a 10 mM solution in DMSO (1 mL) or as a dry powder and is offered with high reported purity for research applications.
  • Acts as a formyl peptide receptor (FPR) antagonist.
  • Inhibits formyl peptide receptor like 1 (FPRL1) at higher concentrations.
  • Available as a 10 mM solution in DMSO (1 mL) or as dry powder formats.
  • High reported purity suitable for biochemical assays (~99.5%).
  • Contains Boc-protected Met-Leu-Phe sequence as the trifluoroacetate salt.
Catalog No. 50-002-48919 Supplier Medchemexpress LLC Supplier No. HY103473A10MM 1ML
Boc-MLF TFA is a Boc-protected tripeptide (Boc-Met-Leu-Phe as the trifluoroacetate salt) used as a formyl peptide receptor (FPR) antagonist for receptor pharmacology and cell signaling studies. It is supplied as a 10 mM solution in DMSO (1 mL) or as a dry powder and is offered with high reported purity for research applications.
  • Acts as a formyl peptide receptor (FPR) antagonist.
  • Inhibits formyl peptide receptor like 1 (FPRL1) at higher concentrations.
  • Available as a 10 mM solution in DMSO (1 mL) or as dry powder formats.
  • High reported purity suitable for biochemical assays (~99.5%).
  • Contains Boc-protected Met-Leu-Phe sequence as the trifluoroacetate salt.