Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boc-Met-OSu, Advanced ChemTech

Catalog No. 01341041
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-341-041 25 g
01-341-040 5 g
01-341-042 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-341-041 Supplier Advanced Chem Tech Supplier No. BM240125G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3845-64-5
Molecular Formula C14H22N2O6S
Molecular Weight (g/mol) 346.40
MDL Number MFCD00042760
InChI Key PCZJWSPKNYONIM-VIFPVBQESA-N
IUPAC Name 2,5-dioxopyrrolidin-1-yl (2S)-2-{[(tert-butoxy)carbonyl]amino}-4-(methylsulfanyl)butanoate
SMILES CSCC[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O

Specifications

Color White/Off-White
Quantity 25 g
Grade Biochemical
Chemical Name or Material Boc-Met-OSu
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.