Learn More
Medchemexpress LLC HY-103473A 50mg Medchemexpress, Boc-MLF (TFA) CAS: Purity:>98%
SDP
Catalog No. 501968116
Encompass_Preferred
Medchemexpress, HY-103473A 50mg Boc-MLF (TFA) CAS: Boc-MLF (TFA) is a peptide, used as a specific formyl peptide receptor (FPR) antagonist, also inhibits the signaling through formyl peptide receptor like 1 (FPRL1) at higher concentrations. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8116 Supplier Medchemexpress LLC Supplier No. HY103473A50MG
Medchemexpress, HY-103473A 50mg Boc-MLF (TFA) CAS: Boc-MLF (TFA) is a peptide, used as a specific formyl peptide receptor (FPR) antagonist, also inhibits the signaling through formyl peptide receptor like 1 (FPRL1) at higher concentrations. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.