Learn More
Medchemexpress LLC Boc-N-Amido-PEG3-azide | 642091-68-7 | C13H26N4O5 | 250 MG
SDP
Catalog No. 5000352850
Encompass_Preferred
Boc-N-Amido-PEG3-azide is a PEG-based PROTAC linker employed in the synthesis of PROTACs. This click chemistry reagent features an azide group that enables participation in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne-containing molecules, as well as strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups. It contributes to the formation of PROTACs, which utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group for click chemistry reactions
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Colorless to light yellow liquid appearance
Catalog No. 50-003-52850 Supplier Medchemexpress LLC Supplier No. 250MGHY140835
Boc-N-Amido-PEG3-azide is a PEG-based PROTAC linker employed in the synthesis of PROTACs. This click chemistry reagent features an azide group that enables participation in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne-containing molecules, as well as strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups. It contributes to the formation of PROTACs, which utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group for click chemistry reactions
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Colorless to light yellow liquid appearance