Learn More
Medchemexpress LLC Boc-N-Amido-PEG4-propargyl | 1219810-90-8 | 99.7% | C16H29NO6 | 100 MG
SDP
Catalog No. 5000355161
Encompass_Preferred
Boc-N-Amido-PEG4-propargyl is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. PROTACs themselves contain two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase, and the other for the target protein, enabling PROTACs to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups
Catalog No. 50-003-55161 Supplier Medchemexpress LLC Supplier No. HY140879100MG
Boc-N-Amido-PEG4-propargyl is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. PROTACs themselves contain two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase, and the other for the target protein, enabling PROTACs to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups