Learn More
Medchemexpress LLC Boc-N-Amido-PEG4-propargyl | 1219810-90-8 | 99.73% | C16H29NO6 | 500 MG
SDP
Catalog No. 5000355162
Encompass_Preferred
Boc-N-Amido-PEG4-propargyl is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is a click chemistry reagent containing an Alkyne group, capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that have Azide groups. PROTACs themselves consist of two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein, and they degrade target proteins by exploiting the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Alkyne group
  • Capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Degrades target proteins by exploiting the intracellular ubiquitin-proteasome system
Catalog No. 50-003-55162 Supplier Medchemexpress LLC Supplier No. HY140879500MG
Boc-N-Amido-PEG4-propargyl is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is a click chemistry reagent containing an Alkyne group, capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that have Azide groups. PROTACs themselves consist of two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein, and they degrade target proteins by exploiting the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Alkyne group
  • Capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Degrades target proteins by exploiting the intracellular ubiquitin-proteasome system