Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-N-ethylglycine | 149794-10-5 | MFCD00672508 | 97.0% | 203.24 g/mol | C9H17NO4 | 10 G
SDP

Catalog No. 5000210582
Encompass_Preferred

Boc-N-ethylglycine is a Boc-protected derivative of N-ethylglycine provided as a research-grade amino acid derivative. It has CAS number 149794-10-5, a molecular weight of 203.24 g/mol, and is typically supplied at ~97.0% purity. The compound is soluble in DMSO and is intended for laboratory research use only.

  • Cas 149794-10-5
  • Molecular formula C9H17NO4
  • Molecular weight 203.24 g/mol
  • Purity 97.0%
  • Soluble in DMSO (≈100 mg/mL); may require ultrasonic agitation
  • Storage: powder -20°C (long term) or 4°C (short term)
  • Pack size: 10 G

Catalog No. 50-002-10582 Supplier Medchemexpress LLC Supplier No. HYW05078210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-N-ethylglycine is a Boc-protected derivative of N-ethylglycine provided as a research-grade amino acid derivative. It has CAS number 149794-10-5, a molecular weight of 203.24 g/mol, and is typically supplied at ~97.0% purity. The compound is soluble in DMSO and is intended for laboratory research use only.

  • Cas 149794-10-5
  • Molecular formula C9H17NO4
  • Molecular weight 203.24 g/mol
  • Purity 97.0%
  • Soluble in DMSO (≈100 mg/mL); may require ultrasonic agitation
  • Storage: powder -20°C (long term) or 4°C (short term)
  • Pack size: 10 G

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.