Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-N-ethylglycine | 149794-10-5 | MFCD00672508 | 97.0% | 203.24 g/mol | C9H17NO4 | 25 G
SDP

Catalog No. 5000209308
Encompass_Preferred

Boc-N-ethylglycine is a Boc-protected N-ethyl derivative of glycine provided as a purified solid for research use. It is used as an amino acid building block in peptide synthesis and other biochemical research applications.

  • Purity 97.0%.
  • Molecular weight 203.24 g/mol.
  • Chemical formula C9H17NO4.
  • Solid form; hygroscopic; soluble in DMSO (100 mg/mL) with ultrasonic assistance.
  • Storage: powder -20°C (3 years) or 4°C (2 years); solutions -80°C (6 months) or -20°C (1 month).
  • Available pack sizes include 10 g, 25 g, 100 g, 500 g.

Catalog No. 50-002-09308 Supplier Medchemexpress LLC Supplier No. 25GHYW050782
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-N-ethylglycine is a Boc-protected N-ethyl derivative of glycine provided as a purified solid for research use. It is used as an amino acid building block in peptide synthesis and other biochemical research applications.

  • Purity 97.0%.
  • Molecular weight 203.24 g/mol.
  • Chemical formula C9H17NO4.
  • Solid form; hygroscopic; soluble in DMSO (100 mg/mL) with ultrasonic assistance.
  • Storage: powder -20°C (3 years) or 4°C (2 years); solutions -80°C (6 months) or -20°C (1 month).
  • Available pack sizes include 10 g, 25 g, 100 g, 500 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.