Learn More
Medchemexpress LLC Boc-N-gamma-trityl-D-asparagin | 210529-01-4 | 98.6% | 474.55 g/mol | C28H30N2O5 | 10 G
SDP
Catalog No. 5000208330
Encompass_Preferred
Boc-N-gamma-trityl-D-asparagin is a Boc- and trityl-protected D-asparagine derivative used as a protected amino acid building block in peptide synthesis and organic synthetic workflows. It provides Boc protection on the amino group and trityl protection on the side-chain amide, enabling orthogonal deprotection strategies and protecting asparagine residues during multi-step syntheses.
  • Provides Boc protection on the α-amino group and trityl protection on the side-chain amide.
  • Suitable for peptide synthesis and orthogonal protection strategies.
  • High purity for research applications.
  • Available in gram-scale packs for laboratory use.
Catalog No. 50-002-08330 Supplier Medchemexpress LLC Supplier No. HYW08810010G
Boc-N-gamma-trityl-D-asparagin is a Boc- and trityl-protected D-asparagine derivative used as a protected amino acid building block in peptide synthesis and organic synthetic workflows. It provides Boc protection on the amino group and trityl protection on the side-chain amide, enabling orthogonal deprotection strategies and protecting asparagine residues during multi-step syntheses.
  • Provides Boc protection on the α-amino group and trityl protection on the side-chain amide.
  • Suitable for peptide synthesis and orthogonal protection strategies.
  • High purity for research applications.
  • Available in gram-scale packs for laboratory use.