Learn More
Medchemexpress LLC Boc-N-PEG1-C2-NHS ester | 1260092-55-4 | 97.0% | C14H22N2O7 | 10 G
SDP
Catalog No. 5000354540
Encompass_Preferred
Boc-N-PEG1-C2-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs (Proteolysis Targeting Chimeras) contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
Catalog No. 50-003-54540 Supplier Medchemexpress LLC Supplier No. HY14111610G
Boc-N-PEG1-C2-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs (Proteolysis Targeting Chimeras) contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs