Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-N-PEG1-C2-NHS ester | 1260092-55-4 | 97.0% | C14H22N2O7 | 5 G
SDP

Catalog No. 5000353184
Encompass_Preferred

Boc-N-PEG1-C2-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is for research use only. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins, containing two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein.

  • Appears as a solid
  • Color ranges from white to off-white
  • Molecular weight is 330.33
  • Suitable for PROTAC synthesis

Catalog No. 50-003-53184 Supplier Medchemexpress LLC Supplier No. HY1411165G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-N-PEG1-C2-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is for research use only. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins, containing two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein.

  • Appears as a solid
  • Color ranges from white to off-white
  • Molecular weight is 330.33
  • Suitable for PROTAC synthesis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.