Learn More
Medchemexpress LLC Boc-N-PEG2-MS | 302331-20-0 | 98.0% | C10H21NO6S | 100 MG
SDP
Catalog No. 5000356192
Encompass_Preferred
Boc-N-PEG2-MS is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds that leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins. They consist of two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Synthesis of PROTACs
Catalog No. 50-003-56192 Supplier Medchemexpress LLC Supplier No. HY140390100MG
Add to Cart
Add to Cart
Boc-N-PEG2-MS is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds that leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins. They consist of two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Synthesis of PROTACs