Learn More
Medchemexpress LLC Boc-N-PEG2-MS | 302331-20-0 | 98.0% | C10H21NO6S | 25 MG
SDP
Catalog No. 5000352673
Encompass_Preferred
Boc-N-PEG2-MS is a PEG-based PROTAC linker designed for the synthesis of PROTACs. These PROTAC molecules function by bringing together an E3 ubiquitin ligase and a target protein through two connected ligands, ultimately leading to the selective degradation of the target protein via the intracellular ubiquitin-proteasome system.
  • PEG-based linker
  • Essential for PROTAC synthesis
  • Enables targeted protein degradation
  • Utilizes the ubiquitin-proteasome system
Catalog No. 50-003-52673 Supplier Medchemexpress LLC Supplier No. 25MGHY140390
Add to Cart
Add to Cart
Boc-N-PEG2-MS is a PEG-based PROTAC linker designed for the synthesis of PROTACs. These PROTAC molecules function by bringing together an E3 ubiquitin ligase and a target protein through two connected ligands, ultimately leading to the selective degradation of the target protein via the intracellular ubiquitin-proteasome system.
  • PEG-based linker
  • Essential for PROTAC synthesis
  • Enables targeted protein degradation
  • Utilizes the ubiquitin-proteasome system