Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-N-PEG2-MS | 302331-20-0 | 98.0% | C10H21NO6S | 50 MG
SDP

Catalog No. 5000356998
Encompass_Preferred

Boc-N-PEG2-MS is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets the protein of interest. This mechanism exploits the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Achieves selective degradation of target proteins
  • Purity of 98.0%
  • Molecular weight of 283.34
  • Chemical formula C10H21NO6S
  • Appearance is liquid
  • Color is colorless to light yellow

Catalog No. 50-003-56998 Supplier Medchemexpress LLC Supplier No. HY14039050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-N-PEG2-MS is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets the protein of interest. This mechanism exploits the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Achieves selective degradation of target proteins
  • Purity of 98.0%
  • Molecular weight of 283.34
  • Chemical formula C10H21NO6S
  • Appearance is liquid
  • Color is colorless to light yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.